missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone H1.0 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109812
This item is not returnable.
View return policy
Description
Immunogen sequence: MTENSTSAPA AKPKRAKASK KSTDHPKYSD MIVAAIQAEK NRAGSSRQSI QKYIKSHYKV GENADSQIKL SIKRLVTTGV LKQTKGVGAS GSFRLAKSDE PKKSVAFKKT KKEIKKVATP KKASKPKKAA SKAPTKKPKA TPVKKAKKKL AATPKKAKKP KTVKAKPVKA SKPKKAKPVK PKAKSSAKRA GKKK.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a replication-independent histone that is a member of the histone H1 family.
Specifications
| Histone H1.0 | |
| Polyclonal | |
| Unconjugated | |
| H1-0 | |
| D130017D06Rik; H1 histone family member 0; H1 histone family, member 0; H1(0); H1.0, H1(0), H1-0; H10; H1-0 histone; H1f0; H1fv; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1-0; MyD196 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14958, 24437, 3005 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07305, P10922, P43278 | |
| H1-0, H1F0 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human H1F0 (NP_005309.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction