missing translation for 'onlineSavingsMsg'
Learn More

Histone Deacetylase 4/HDAC4, p Ser632 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18407882 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407882 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407882 Supplier Novus Biologicals Supplier No. NBP254636AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histone Deacetylase 4/HDAC4 Polyclonal antibody specifically detects Histone Deacetylase 4/HDAC4 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone Deacetylase 4/HDAC4
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3
Host Species Rabbit
Immunogen HDAC4 (Ab-632) antibody was raised against a peptide sequence around aa. 630∼634 (A-Q-S-S-P) derived from Human HDAC4.
Purification Method Epitope affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 9759
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.