missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Antibody [Allophycocyanin], Novus Biologicals™
Shop All Bio Techne ProductsDescription
Histone Deacetylase 2/HDAC2 Polyclonal antibody specifically detects Histone Deacetylase 2/HDAC2 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Histone Deacetylase 2/HDAC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 |
| Host Species | Sheep |
| Immunogen | E. coli-derived recombinant human Histone Deacetylase 2/HDAC2, Pro386-Pro488, Accession # Q92769 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?