missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-44095AF488
This item is not returnable.
View return policy
Description
Histone Deacetylase 2/HDAC2 Polyclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Histone Deacetylase 2/HDAC2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 | |
| Histone Deacetylase 2/Histone Deacetylase 2/HDAC2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the near C-terminal region of human Histone Deacetylase 2/HDAC2. (Uniprot: Q92769) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Epigenetics, Phospho Specific, Stem Cell Markers | |
| 3066 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction