missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Histone Core Polyclonal Antibody
GREENER_CHOICE

Product Code. 11565843
Change view
Click to view available options
Quantity:
125 μg
Unit Size:
125µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11565843 125 μg 125µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11565843 Supplier Invitrogen™ Supplier No. PA128375

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The histone core is an integral component of chromatin structure, consisting of an octamer of core histones: H2A, H2B, H3, and H4. Each nucleosome unit contains two copies of each of these histones, forming a structural foundation around which approximately 147 base pairs of DNA are wrapped. The histone octamer is organized with an (H3-H4)2 tetramer at its core, flanked by two H2A-H2B dimers. This highly conserved structure not only facilitates DNA compaction but also plays a critical role in regulating gene expression through various post-translational modifications, such as acetylation, methylation, and phosphorylation. These modifications can alter chromatin dynamics and accessibility, impacting processes such as transcription, replication, and DNA repair. The presence of specific histone variants further adds to the epigenetic complexity and regulation of the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone Core
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2.42 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene H4C2
Gene Accession No. P62803, Q5E9F8
Gene Alias core histone macro-H2A.1; H2A histone family, member Y; H2AFY; H2B; H2B clustered histone 19; H2B clustered histone 8; H2BC19; H2BC8; H3 histone, family 3A; H3.3 histone A; H3-3A; H3-3B; H3F3A; H3F3B; H4; H4 clustered histone 2; H4.1; H4C2; histone H2B; histone H2B type 1; histone H2B type 2-E; histone H2B-like; histone H3.3; Histone H4; histone H4.1; macroH2A.1 histone; MACROH2A1; similar to H2A histone family, member Y isoform 2; similar to H3 histone, family 3A
Gene Symbols H2B, H3-3A, H4C2, MACROH2A1
Host Species Sheep
Immunogen Intact calf histone core - rRNA complex.
Purification Method Protein G
Quantity 125 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280691, 326601, 519934, 532990, 615043, 787581
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.