missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone 1F0 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550984
This item is not returnable.
View return policy
Description
Histone H1.0 / H1F0 Polyclonal Antibody detects endogenous levels of Histone H1.0 / H1F0 protein.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a replication-independent histone that is a member of the histone H1 family.
Specifications
| Histone 1F0 | |
| Polyclonal | |
| Unconjugated | |
| H1-0 | |
| D130017D06Rik; H1 histone family member 0; H1 histone family, member 0; H1(0); H1.0, H1(0), H1-0; H10; H1-0 histone; H1f0; H1fv; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1-0; MyD196 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3005 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| P07305 | |
| H1-0, H1F0 | |
| Synthesized peptide derived from the Internal region of human Histone 1.0 (aa 71-120). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction