missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Histochemical Substrate

Product Code. 12871600
Click to view available options
Quantity:
100mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 12871600

Brand: Fisher BioReagents™ 12871600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Application) β-D-Glucosidase
Quantity 100mg
Product Type Histochemical Substrate
Storage Requirements 2 to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.