missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histidase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-53135
This item is not returnable.
View return policy
Description
Histidase Polyclonal specifically detects Histidase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Histidase | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 4.3.1, EC 4.3.1.3, HIS, Histidase, histidine ammonia-lyase, HSTD | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3034 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1.25 ug/ml | |
| P42357 | |
| HAL | |
| Synthetic peptides corresponding to HAL(histidine ammonia-lyase) The peptide sequence was selected from the N terminal of HAL. Peptide sequence INKLQELQVNLVRSHSSGVGKPLSPERCRMLLALRINVLAKGYSGISLET. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Rabbit: 100%; Xenopus: 92%; Canine: 92%; Pig: 92%; Chicken: 85%; Zebrafish: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction