missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine N-Methyltransferase/HNMT Rabbit anti-Human, Clone: 34, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90171-100UL
This item is not returnable.
View return policy
Description
Histamine N-Methyltransferase/HNMT Monoclonal specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| Histamine N-Methyltransferase/HNMT | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3176 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| 34 | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 ul/mg of lysate | |
| EC 2.1.1.8, histamine N-methyltransferase, HMT, HNMT-S1, HNMT-S2 | |
| Recombinant Human Histamine N-Methyltransferase/HNMT protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction