Learn More
Histamine N-Methyltransferase/HNMT Antibody, Janelia Fluor™ 549, Novus Biologicals™
Product Code. 18415719 Shop All Bio Techne Products
Product Code. 18415719 Supplier Novus Biologicals Supplier No. NBP300070JF549
Rabbit Polyclonal Antibody
Histamine N-Methyltransferase/HNMT Polyclonal antibody specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation