missing translation for 'onlineSavingsMsg'
Learn More

Histamine N-Methyltransferase/HNMT Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30733343 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733343 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733343 Supplier Novus Biologicals Supplier No. AF7637CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Histamine N-Methyltransferase/HNMT Polyclonal antibody specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histamine N-Methyltransferase/HNMT
Applications Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias EC 2.1.1.8, histamine N-methyltransferase, HMT, HNMT-S1, HNMT-S2
Host Species Sheep
Immunogen E. coli-derived recombinant human Histamine N-Methyltransferase/HNMT, Ser7-Ala292, Accession # P50135
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 3176
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.