missing translation for 'onlineSavingsMsg'
Learn More

Histamine N-Methyltransferase/HNMT Antibody, Biotin, Novus Biologicals™

Product Code. 18344294 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344294 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344294 Supplier Novus Biologicals Supplier No. NBP300070B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histamine N-Methyltransferase/HNMT Polyclonal specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histamine N-Methyltransferase/HNMT
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias EC 2.1.1.8, histamine N-methyltransferase, HMT, HNMT-S1, HNMT-S2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Histamine N-Methyltransferase/HNMT (AAH20677.1; Met 1-Ala 292).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 3176
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.