missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine N-Methyltransferase/HNMT Antibody (034), Janelia Fluor™ 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Histamine N-Methyltransferase/HNMT Monoclonal antibody specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Classification | Monoclonal |
| Clone | 034 |
| Formulation | 50mM Sodium Borate |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?