missing translation for 'onlineSavingsMsg'
Learn More

Histamine H4R Antibody - BSA Free, Novus Biologicals™

Product Code. 18321654 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321654 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321654 Supplier Novus Biologicals Supplier No. NLS1901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histamine H4R Polyclonal specifically detects Histamine H4R in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histamine H4R
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 3.6 ug/ml
Formulation PBS with 0.1% Sodium Azide
Gene Alias AXOR35H4, GPCR105MGC133027, G-protein coupled receptor 105, GPRv53histamine H4 receptor, H4Rpfi-013, HH4RBG26, histamine receptor H4, Pfi-013, SP9144
Gene Symbols HRH4
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 2nd extracellular domain of human Histamine H4R.
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 59340
Test Specificity Histamine H4 Receptor
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.