missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Histamine H4R Polyclonal antibody specifically detects Histamine H4R in Human, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Histamine H4R |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 20 μg/mL |
| Formulation | PBS |
| Gene Accession No. | Q9H3N8 |
| Gene Alias | AXOR35H4, GPCR105MGC133027, G-protein coupled receptor 105, GPRv53histamine H4 receptor, H4Rpfi-013, HH4RBG26, histamine receptor H4, Pfi-013, SP9144 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?