missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine H3R Antibody (27H8), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Histamine H3R |
|---|---|
| Clone | 27H8 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30027867
|
Novus Biologicals
NBP3-26219-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30029262
|
Novus Biologicals
NBP3-26219-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histamine H3R Monoclonal antibody specifically detects Histamine H3R in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Histamine H3R | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, histamine receptor H3 | |
| A synthesized peptide derived from Human Histamine H3R [UniProt Q9Y5N1] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 27H8 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| GPCR, Immunology, Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 11255 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title