missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine H1R Antibody (480054), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4726H
365.50 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Histamine H1R Monoclonal antibody specifically detects Histamine H1R in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Histamine H1R | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 480054 | |
| Flow Cytometry, CyTOF-ready | |
| H1R, H1-R, hisH1, histamine receptor H1, subclass H1 | |
| NS0 mouse myeloma cell line transfected with human Histamine H1 R, Met1-Ser487, Accession # AAH60802 | |
| 0.1 mL | |
| GPCR, Immune System Diseases, Immunology, Innate Immunity | |
| 3269 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction