missing translation for 'onlineSavingsMsg'
Learn More

Histamine Diphosphate, TRC

Product Code. 30392618
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392618 1 g -
30405631 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392618 Supplier Toronto Research Chemicals Supplier No. TRCH4560601G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-74-1
Molecular Formula C5 H9 N3 . 2 H3 O4 P
Molecular Weight (g/mol) 307.14
Synonym Histamine, diphosphate (7CI), Histamine, phosphate (1:2) (6CI,8CI), 4-(2-Aminoethyl)imidazole bis(dihydrogen phosphate), 4-(2-Aminoethyl)imidazole di-acid phosphate, Histamine acid phosphate, Histamine biphosphate, Histamine dihydrogen phosphate, Histamine phosphate, Histapon, 1H-Imidazole-4-ethanamine, phosphate (1:2), Histamine Phosphate
IUPAC Name 2-(1H-imidazol-4-yl)ethanamine;phosphoric acid
SMILES NCCc1c[nH]cn1.OP(=O)(O)O.OP(=O)(O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H9N3.2H3O4P/c6-2-1-5-3-7-4-8-5;2*1-5(2,3)4/h3-4H,1-2,6H2,(H,7,8);2*(H3,1,2,3,4)
Formula Weight 307.0334
Chemical Name or Material Histamine Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.