missing translation for 'onlineSavingsMsg'
Learn More

Histamine diphosphate, 98%

Product Code. 15493739
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493739 5 g 5g
15403749 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493739 Supplier Thermo Scientific Alfa Aesar Supplier No. J61088.06
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Endogenous H1 and H2 agonist | CAS: 51-74-1 | C5H15N3O8P2 | 307.14 g/mol

Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-74-1
Molecular Formula C5H15N3O8P2
Molecular Weight (g/mol) 307.14
MDL Number MFCD00064316
InChI Key ZHIBQGJKHVBLJJ-UHFFFAOYSA-N
Synonym histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate
PubChem CID 65513
ChEBI CHEBI:51193
SMILES OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1

Specifications

Melting Point 128°C to 132°C
Color White
Quantity 5 g
Sensitivity Hygroscopic
Merck Index 14,4719
Solubility Information Soluble in water.
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine;phosphoric acid
Formula Weight 307.14
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Histamine diphosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.