missing translation for 'onlineSavingsMsg'
Learn More

Histamine dihydrochloride, 99%

Product Code. 11996251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11996251 500 g Glass bottle 500g
10583311 5 g Glass bottle 5g
10593501 25 g Glass bottle 25g
10503511 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11996251 Supplier Thermo Scientific Acros Supplier No. 150625000
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 56-92-8 | C5H11Cl2N3 | 184.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-92-8
Molecular Formula C5H11Cl2N3
Molecular Weight (g/mol) 184.06
MDL Number MFCD00012703
InChI Key PPZMYIBUHIPZOS-UHFFFAOYSA-N
Synonym histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
PubChem CID 5818
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1

Specifications

Melting Point 249.0°C to 252.0°C
Color White
Loss on Drying 0.5% (105°C) max.
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Quantity 500 g
Beilstein 25315
Merck Index 154756
Solubility Information Solubility in water: freely soluble in water. Other solubilities: soluble in ethanol,freely soluble in methanol
Formula Weight 184.07
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Histamine dihydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Histamine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.