missing translation for 'onlineSavingsMsg'
Learn More

Histamine dihydrochloride, 98+%

Product Code. 11387276
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387276 5 g 5g
11397276 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387276 Supplier Thermo Scientific Alfa Aesar Supplier No. L09198.06
  • ONLINE EXCLUSIVE PRICE
    £43.35
    £37.65 / 5g
    Save £5.70
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-92-8 | C5H11Cl2N3 | 184.06 g/mol

Histamine dihydrochloride has been shown to activate nitric oxide synthetase and suppress or inhibit the generation of reactive oxygen species (ROS). Inhibition of ROS by histamine dihydrochloride allows activation of T cells and NK cells by IL-2. In a rat model, histamine dihydrochloride suppressed ROS generated by Kupffer cells through the H2 histamine receptor. It is a potent vasodilator and endogenous histamine receptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histamine dihydrochloride has been shown to activate nitric oxide synthetase and suppress or inhibit the generation of reactive oxygen species (ROS). Inhibition of ROS by histamine dihydrochloride allows activation of T cells and NK cells by IL-2. In a rat model, histamine dihydrochloride suppressed ROS generated by Kupffer cells through the H2 histamine receptor. It is a potent vasodilator and endogenous histamine receptor agonist.

Solubility
Very soluble in water and methanol. Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-92-8
Molecular Formula C5H11Cl2N3
Molecular Weight (g/mol) 184.06
MDL Number MFCD00012703
InChI Key PPZMYIBUHIPZOS-UHFFFAOYSA-N
Synonym histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
PubChem CID 5818
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1

Specifications

Melting Point 245°C to 249°C
Quantity 5 g
Beilstein 3624116
Merck Index 14,4719
Solubility Information Very soluble in water and methanol. Soluble in ethanol.
Formula Weight 184.07
Percent Purity ≥98%
Chemical Name or Material Histamine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.