missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Histamine Competitive ELISA Kit

Product Code. 18097414
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18097414

Brand: Invitrogen™ EEL005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Histamine is located in mast cells, endocrine cells of the gut, blood cells and in some cells of the peripheral and central nervous system. Histamine is a potent vasodilator when secreted by mast cell found in various tissues as a result of allergic hytersensitivity or inflammation. In the central nervous system, Histamine is putative neurotransmitter. In the brain, its highest content has been found in the hypothalamus and in certain areas of the mesencephalon. The Histamine antiserum has a sensitivity level capable of detecting the low level Histamine contents of the brain.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.56 to 100 ng/mL
Assay Sensitivity 0.94 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Histamine
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.