missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST2H2BE, Mouse anti-Human, Clone: 4G6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008349-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: ESYSIYVYKVLKQVHPDTGISSKAMGIMNSFVNDIFERIAGEASRLAHYNKRSTITSREIQTAVRLLLPGELAKHAVSEGTKAVTKYTSSKSpecifications
| HIST2H2BE | |
| Monoclonal | |
| Unconjugated | |
| histone cluster 2, H2be | |
| GL105/H2B/H2B.1/H2B/q/H2BFQ/MGC119802/MGC119804/MGC129733/MGC129734 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin) | |
| 4G6 | |
| In 1x PBS, pH 7.4 | |
| NM_003528 | |
| HIST2H2BE | |
| HIST2H2BE (NP_003519.1, 36 a.a. ∼ 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8349 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction