missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIST2H2AA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585584
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Rat (100%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.
Specifications
| HIST2H2AA3 | |
| Polyclonal | |
| Unconjugated | |
| H2AC18 | |
| H2A; H2A histone family, member O; H2A.2; H2A/O; H2A/q; H2a-615; H2AFO; HIST2H2AA; Hist2h2aa1; HIST2H2AA3; HIST2H2AA4; histone 2, H2aa; histone 2, H2aa3; histone cluster 2, H2aa; histone cluster 2, H2aa1; histone cluster 2, H2aa3; histone H2A type 2-A; Histone H2A.2; Histone H2A/o | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8337 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6FI13 | |
| H2AC18 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Histone H2A.2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction