missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2BC Antibody (CL5101), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61154-25ul
This item is not returnable.
View return policy
Description
HIST1H2BC Monoclonal specifically detects HIST1H2BC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HIST1H2BC | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| HIST1H2BC | |
| This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:VSEGTKAVTKYTSSK | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL5101 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| H2B histone family, member K, H2B/a, H2B/g, H2B/kH2B/h, H2BFA, H2BFG, H2BFH, H2BFKH2B/l, H2BFL, HIST1H2BC, HIST1H2BE, HIST1H2BF, HIST1H2BG, histone 1, H2bi, histone cluster 1, H2bi, histone H2B type 1-C/E/F/G/I, Histone H2B.1 A, Histone H2B.a, Histone H2B.g, Histone H2B.h, Histone H2B.k, Histone H2B.l | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8347 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction