missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIST1H2BB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117474
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
| HIST1H2BB | |
| Polyclonal | |
| Unconjugated | |
| H2BC3 | |
| H2B histone family, member F; H2B.1; H2B/f; h2B-143; H2B-clustered histone 3; H2BFF; HIST1H2BB; histone 1, H2bb; histone cluster 1, H2bb; histone H2B type 1-B; Histone H2B.1; histone H2B.f | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3018, 319178 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P33778, Q64475 | |
| H2BC3, HIST1H2BB | |
| Peptide sequence around site of Lys (16) derived from Human Histone H2B type 1-B. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction