missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIST1H2AG Monoclonal Antibody (CL5041)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA535025
This item is not returnable.
View return policy
Description
Immunogen sequence: RSSRAGLQFP VGRVHR Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Histones are nuclear proteins that play an important role in maintaining the structure of the chromosome in eukaryotes. The nucleosome is made up of around 146bp of DNA, which enfolds around a histone octomer unit comprising of four major histones, H2A, H2B, H3 and H4. An important role of H2A is to ensure proper assembly or functioning of the centrometre. They also play an important role in gene regulation. The protein can be modified by acetylation and phosphorylation.
Specifications
| HIST1H2AG | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0C0S8 | |
| H2AC11, HIST1H2AG | |
| Synthetic Peptide corresponding to Human HIST1H2AG. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| CL5041 | |
| Unconjugated | |
| H2AC11 | |
| H2A histone family, member P; H2A.1; H2A.1b; H2A/d; H2A/p; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; H2AFC; H2AFD; H2AFI; H2AFN; H2AFP; H2AG; H2ak; HIST1H2AG; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; histone 1, H2ag; histone cluster 1, H2ag; histone H2A type 1; histone H2A/e; histone H2A/p; Histone H2A/ptl; pH2A/f | |
| Mouse | |
| Protein A | |
| RUO | |
| 8329, 8969 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction