missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2AG Antibody (CL5039), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61151
This item is not returnable.
View return policy
Description
HIST1H2AG Monoclonal specifically detects HIST1H2AG in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HIST1H2AG | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| HIST1H2AG | |
| This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:RSSRAGLQFPVGRVHR | |
| 100 μL | |
| 8969 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CL5039 | |
| Western Blot 1 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| H2A histone family, member P, H2A.1, H2A.1b, H2A/p, H2AFC, H2AFD, H2AFI, H2AFN, H2AFP, HIST1H2AI, HIST1H2AK, HIST1H2AL, HIST1H2AM, histone 1, H2ag, histone cluster 1, H2ag, histone H2A type 1, Histone H2A/p, pH2A/f | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction