missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2AG Antibody (CL5039) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61151JF525
This item is not returnable.
View return policy
Description
HIST1H2AG Monoclonal antibody specifically detects HIST1H2AG in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| HIST1H2AG | |
| Monoclonal | |
| Janelia Fluor 525 | |
| H2A histone family, member P, H2A.1, H2A.1b, H2A/p, H2AFC, H2AFD, H2AFI, H2AFN, H2AFP, HIST1H2AI, HIST1H2AK, HIST1H2AL, HIST1H2AM, histone 1, H2ag, histone cluster 1, H2ag, histone H2A type 1, Histone H2A/p, pH2A/f | |
| This antibody was developed using a synthetic peptide derived from P0C0S8, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL5039 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8969 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction