missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HISPPD2A Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44293CL1
This item is not returnable.
View return policy
Description
HISPPD2A Polyclonal antibody specifically detects HISPPD2A in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| HISPPD2A | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9677 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| diphosphoinositol pentakisphosphate kinase 1, histidine acid phosphatase domain-containing protein 2A, inositol pyrophosphate synthase 1, VIP1 | |
| The epitope recognized by this antibody maps to a region between residue 1383 to 1433 of human Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1 using the numbering given in entry Q6PFW1.1 (GeneID 9677). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction