missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HISPPD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | HISPPD1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667652
|
Novus Biologicals
NBP2-98035-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619764
|
Novus Biologicals
NBP2-98035-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HISPPD1 Polyclonal antibody specifically detects HISPPD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| HISPPD1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| PBS (pH 7.0) | |
| 23262 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| diphosphoinositol pentakisphosphate kinase 2FLJ21506, EC 2.7.4.21, EC 2.7.4.24, HISPPD1histidine acid phosphatase domain containing 1, Histidine acid phosphatase domain-containing protein 1, hsVIP2, inositol heptaphosphate kinase 2, inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2, InsP6 and PP-IP5 kinase 2, KIAA0433FLJ23463, VIP1 homolog 2, VIP2IP7K2 | |
| Produced in rabbits immunized with E. coli-derived Human HISPPD1 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title