missing translation for 'onlineSavingsMsg'
Learn More

Hispidulin, TRC

Product Code. 30389573
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389573 25 mg
30389042 5 mg
30403840 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389573 Supplier Toronto Research Chemicals Supplier No. TRCH45634025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1447-88-7
Molecular Formula C16H12O6
Molecular Weight (g/mol) 300.27
Synonym 5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 4',5,7-Trihydroxy-6-methoxyflavone, 5,7,4'-Trihydroxy-6-methoxyflavone, 6-Methoxy-5,7,4'-trihydroxyflavone, 6-Methoxyapigenin, 6-Methylscutellarein, 6-O-Methylscutellarein, Dinatin, Hispedulin, Hispidulin, Hispiludin, NSC 122415, Salvitin, Scutellarein 6-Methyl Ether, 4',5,7-Trihydroxy-6-methoxy-flavone
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)-6-methoxychromen-4-one
SMILES COc1c(O)cc2OC(=CC(=O)c2c1O)c3ccc(O)cc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H12O6/c1-21-16-11(19)7-13-14(15(16)20)10(18)6-12(22-13)8-2-4-9(17)5-3-8/h2-7,17,19-20H,1H3
Formula Weight 300.0634
Chemical Name or Material Hispidulin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.