missing translation for 'onlineSavingsMsg'
Learn More

Hispidulin, MedChemExpress

Product Code. 16439248
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16439248

Brand: Medchem Express HYN1950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hispidulin is a natural flavone with a broad spectrum of biological activities. Hispidulin is a Pim-1 inhibitor with an IC50 of 2.71 μM.

Hispidulin is a natural flavone with a broad spectrum of biological activities. Hispidulin is a Pim-1 inhibitor with an IC50 of 2.71 μM.

Specifications

Chemical Name or Material Hispidulin
CAS 1447-88-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H12O6
Quantity 10 mg
Synonym Dinatin
Solubility Information DMSO : 62.5 mg/mL (208.15 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(O)C=C2)OC3=CC(O)=C(OC)C(O)=C13
Molecular Weight (g/mol) 300.26
Formula Weight 300.26
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.