missing translation for 'onlineSavingsMsg'
Learn More

Hispidulin, MedChemExpress

Product Code. 16481658
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481658 5 mg 5mg
16453936 25 mg 25mg
16439248 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481658 Supplier Medchem Express Supplier No. HYN19505mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hispidulin is a natural flavone with a broad spectrum of biological activities. Hispidulin is a Pim-1 inhibitor with an IC50 of 2.71 μM.

Hispidulin is a natural flavone with a broad spectrum of biological activities. Hispidulin is a Pim-1 inhibitor with an IC50 of 2.71 μM.

Specifications

Chemical Name or Material Hispidulin
CAS 1447-88-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H12O6
Quantity 5 mg
Synonym Dinatin
Solubility Information DMSO : 62.5 mg/mL (208.15 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(O)C=C2)OC3=CC(O)=C(OC)C(O)=C13
Molecular Weight (g/mol) 300.26
Formula Weight 300.26
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.