missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HisG Epitope Tag Monoclonal Antibody, HRP

Product Code. 10464752
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10464752 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 10464752 Supplier Invitrogen™ Supplier No. R94125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

Mouse Monoclonal Antibody

NOTE: R941-25 was previously sold under product #46-1009. When re-ordering, please use #R941-25. This antibody is designed to specifically detect recombinant proteins containing the seven amino acid 6xHis-Gly epitope. This epitope is present in many expression vectors encoding an N-terminal 6xHis tag. The HisG antibody recognizes the following sequence: -His-His-His-His-His-His-Gly and can be used to detect expression of fusion proteins from bacterial, insect, and mammalian cells. Anti-HisG-HRP Antibody was prepared by crosslinking the Anti-HisG Antibody with horseradish peroxidase using glutaraldehyde. It has been tested in immunoblotting and ELISA procedures. It was tested against purified Positope™ control protein (5 ng). The Positope™ control protein is a 53 kDa recombinant protein that contains seven epitope tags, including His(C-term), HisG, c-Myc, and V5. Low background was observed using chemiluminescent or alkaline phosphatase reagents for detection. In western blot experiments with purified protein, 50 ng (for Anti-HisG-HRP Antibody) of the Positope protein gave a detectable signal. For western blot, dilute in PBS containing 0.05% Tween-20 and 5% nonfat, dry milk (PBSTM). Using chemiluminescence as the detection method, no cross-reactivity has been observed in bacterial lysates. In mammalian lysates, a few cross-reactive proteins have been observed upon overexposure of blots. If you use alkaline phosphatase-conjugated secondary antibody, do not use PB...

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Expression of recombinant proteins in E. coli as a fusion protein with neighboring histidine residues is one of the most popular methods of epitope tags. The affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. HisG antibodies allow detection of recombinant proteins containing a polyhistidine sequence: His-His-His-His-His-His-Gly (6xHis-Gly epitope). HisG antibodies can be used to detect expression of fusion proteins from bacterial, insect, and mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HisG Epitope Tag
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate HRP
Formulation PBS with no preservative
Gene Alias 6xHis-Gly; 6xHis-Glycine; His G; His Tag; Histidine Tag
Host Species Mouse
Immunogen 6x His synthetic peptide: -His-His-His-His-His-His-Gly.
Purification Method Purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.