missing translation for 'onlineSavingsMsg'
Learn More

His-Tag Mouse anti-Tag, HRP, Clone: 1B7G5, Proteintech

Product Code. 16871246 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16871246 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16871246 Supplier Proteintech Supplier No. HRP66005100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody is conjugated with HRP. Note: Do not add azide (Sodium Azide or Smite) into the dilution buffer. azide is the HRP inhibitor which decreases the enzymatic activity of HRP.

The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombit proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombit protein using genetic engineering. By choosing a particular epitope and recombit protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombit proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent.

Specifications

Antigen His-Tag
Applications Western Blot
Classification Monoclonal
Clone 1B7G5
Concentration 0.4 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and 0.02% thimerosal; pH 7.3
Gene Alias 6x His, 6xHis, His Tag, 6histag, 6XHis, HHHHHH, His tag, HISTIDINE TAGGED, HRP 6XHis, HRP HHHHHH, HRP His tag
Host Species Mouse
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.