missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Single-use spin columns prepacked with Ni Sepharose High Performance Medium that is precharged with nickel ions; negligible ion leakage
- One run in standard microcentrifuge takes approximatedly 10 min.
- Yields up to 750μg purified protein per column
- Allows direct purification of both clarified and unclarifed cell lysates
- Also useful for screening large numbers of small-scale bacterial lysates
- Kit option contains buffer, allowing fast and convenient purifications without time-consuming buffer preparation step
- Kit components (spin columns and buffer) can also be purchased separately
Medium Characteristics:
- Compatible with a wide range of reducing agents, detergents, denaturants and other additives
- Stable (long-term) in the pH range 3-12
- Average particle size: 34μm
Order Info
Kit (28-9321-71) contains 50 columns and one pack of His Buffer Kit (2 x 100mL phosphate 8X stock solution, pH 7.4, and 100mL highly pure 2M imidazole, pH 7.4)
Specifications
Specifications
| Kit Contents | 1 Pk. His SpinTrap (28-4013-53) and 1 Pk. His Buffer Kit (11-0034-00) |
| pH Range | 2 to 14 |
| Support Type | 6% Highly Cross-linked Agarose |
| Content And Storage | 4°C to 30°C |
| Quantity | 1 Piece |
| Type | Spin Column Kit |
Safety and Handling
Signal Word
- Danger||
- Skin sensitiser Category 1
- Skin corrosion/irritation Category 1 C
- Reproductive toxicity Category 1B
- H317-May cause an allergic skin reaction.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P331-Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P305-IF IN EYES:Immediately call a POISON CENTER/doctor..
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Nickel, imidazole
- UNKNOWN AQUATIC-Contains 15% of components with unknown hazards to the aquatic environment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?