missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIRA Rabbit anti-Human, Clone: 6Q7Y6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | HIRA |
|---|---|
| Clone | 6Q7Y6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319495
|
Novus Biologicals
NBP3-16716-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18387865
|
Novus Biologicals
NBP3-16716-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIRA Monoclonal antibody specifically detects HIRA in Human samples. It is validated for Western BlotSpecifications
| HIRA | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Chromatin Modifiers, Stem Cells | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 7290 | |
| IgG | |
| Affinity purified |
| 6Q7Y6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DGCR1DiGeorge critical region gene 1, HIR, HIR (histone cell cycle regulation defective) homolog A (S. cerevisiae), HIR histone cell cycle regulation defective homolog A (S. cerevisiae), TUP1, TUP1-like enhancer of split protein 1, TUPLE1protein HIRA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 550-675 of human HIRA (P54198). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title