missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIRA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565749
This item is not returnable.
View return policy
Description
Immunogen sequence: TWNLVSDKQDS LAQCADFRSS LPSQDAMLCS GPLAIIQGRT SNSGRQAARL FSVPHVVQQE TTLAYLENQV AAALTLQSSH EYRHWLLVYA RYLVNEGFEY RLREICKDLL GPVHYSTG Highest antigen sequence identity to the following orthologs - mouse 97%, rat 95%.
HIRA is a histone chaperone that preferentially places the variant histone H3.3 in nucleosomes. Orthologs of this protein in yeast, flies, and plants are necessary for the formation of transcriptionally silent heterochromatin. It plays an important role in the formation of the senescence-associated heterochromatin foci. These foci likely mediate the irreversible cell cycle changes that occur in senescent cells. It is considered the primary candidate protein in some haploinsufficiency syndromes such as DiGeorge syndrome, and insufficient production of the gene may disrupt normal embryonic development.
Specifications
| HIRA | |
| Polyclonal | |
| Unconjugated | |
| HIRA | |
| AA138857; D16Ertd95e; DGCR1; DiGeorge critical region gene 1; Gm15797; HIR; HIR histone cell cycle regulation defective homolog A; Hira; histone cell cycle regulation defective homolog A; histone cell cycle regulation defective homolog A (S. cerevisiae); histone cell cycle regulator; N28177; OTTMUSG00000026264; Protein HIRA; TUP1; TUP1-like enhancer of split gene 1; TUP1-like enhancer of split protein 1; TUPLE1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7290 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54198 | |
| HIRA | |
| Recombinant Human HIRA. Recombinant protein control fragment (Product #RP-106426). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction