missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ Sephacryl™ HR Prepacked Gel Filtration Columns

Product Code. 11350292 Shop All Cytiva Products
Change view
Click to view available options
Flow Rate:
<0.5 mL/min.
<1.3 mL/min.
Bed Volume:
120 mL
320 mL
Diameter (Metric) Inner:
1.6cm
2.6cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Bed Volume Diameter (Metric) Inner unitSize
11350292 <1.3 mL/min. 120 mL 1.6cm Each
11330292 <0.5 mL/min. 120 mL 1.6cm Each
11360292 <0.5 mL/min. 320 mL 2.6cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11350292 Supplier Cytiva Supplier No. 28935606
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

For reproducible protein purification, separating polysaccharides, macromolecules with extended structures and small plasmids

Aids in preparative separation of biomolecules from up to 5mL (for 16mm column I.D.) and 13mL (for 26mm column I.D.).

  • Provides maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability
  • Prepacked columns for use with AKTAdesign™ or other pumped chromatography systems
  • Available in two different prepacked column sizes - 16 x 600mm (120mL) and 26 x 60mm (320mL)
  • Maximum pressure (over packed bed during operation) is 1.5 bar (22psi)

Media    

  • Average particle size: 50μm
  • Matrix: Allyl dextran and N, N'-methylenebisacrylamide
  • pH Stability working range: 3 to 11
  • pH Stability cleaning range: 2 to 13
  • Chemical stability: all commonly used buffers, 0.2 M NaOH, 0.1 M HCl, 1 M acetic acid, 8 M urea, 6 M guanidine HCl, 1% SDS, 20% ethanol, 30% propanol, 30% acetonitrile (tested at 40°C for one week), 0.5 M NaOH (only for cleaning-in-place)
  • Pressure/Flow specification: >125cm/hr., 100kPa, XK 50/30, bed height 15cm
Column
  • Material (bed support): Polymethyl-Methacrylate

HiPrep 16/60 Sephacryl S-400 HR

  • Media:
  • Fractionation [Mr] globular proteins: 2 x 104-8 x 106
  • Exclusion limit [Mr] (globular proteins): ≈9 x 106 (9000kDa)
  • Fractionation range [Mr] DNA exclusion limit:    271bp
  • Fractionation [Mr] Dextrans: 1 x 104-2 x 106
HiPrep 16/60 Sephacryl S-500 HR
  • Media:
  • Exclusion limit for Dextrans: 100 x 106
  • Fractionation range [Mr] DNA exclusion limit: 1078bp
  • Fractionation [Mr] Dextrans: 4 x 104-2 x 107

TRUSTED_SUSTAINABILITY

Specifications

Quantity Each
Flow Rate <1.3 mL/min.
Particle Size 25 to 75 μm
For Use With (Equipment) Low to medium pressure AKTA™ systems or pumped chromatography systems
Bed Volume 120 mL
Product Type Sephacryl HR Pre-packed Gel Filtration Column
Diameter (Metric) Inner 1.6cm
For Use With (Application) Reproducible Protein purification, separating Polysaccharides, macromolecules with extended structures and small plasmids
Height (English) 23.62 in.
Height (Metric) 60cm
Material Polypropylene
Max. Pressure 1.5 bar [0.15 MPa] (22 psi)
pH Range 3 to 11
Support Type Allyl Dextran and N,N'-methylenebisacrylamide
Sample Type Polysaccharides, macromolecules with extended structures and even small plasmids
Content And Storage 4°C to 30°C, 20% Ethanol
Show More Show Less
compliance-icons
Product Identifier
  • HiPrep 16/60 Sephacryl S-500 HR
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.