missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ Q XL 16/10 Column

Product Code. 10489764 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
20 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10489764 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10489764 Supplier Cytiva Supplier No. 28936538
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Provide fast, preparative separations of proteins and other biomolecules

  • Strong anion exchanger prepacked column for initial capture of proteins and other biomolecules with IEX chromatography
  • Compatible with simple pump-based configurations and other chromatography systems such as AKTA design
  • Optimized for reliable, reproducible separations
  • High loading capacity and fast purifications
  • Rapid initial capture of proteins from start material

TRUSTED_SUSTAINABILITY

Specifications

Description HiPrep Q XL 16/10
Diameter (English) Inner Column 0.6 in.
Flow Rate 2 to 10 mL/min.
For Use With (Application) HiPrep Q XL 16/10 is a strong anion exchanger prepacked with Q Sepharose XL columns for initial capture of proteins and other biomoleculs with ion exchange (IEX) chromatography.
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 20 mL
Quantity 20 mL
compliance-icons
Product Identifier
  • HiPrep Q XL 16/10
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.