missing translation for 'onlineSavingsMsg'
Learn More

Hippuric Acid Sodium Salt, TRC

Product Code. 30395784
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395784 500 mg -
30402611 250 mg -
30399671 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395784 Supplier Toronto Research Chemicals Supplier No. TRCH347013500MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-94-5
Molecular Formula C9H8NNaO3
Molecular Weight (g/mol) 201.15
Synonym N-Benzoylglycine Sodium Salt, Sodium Hippurate
IUPAC Name sodium;2-benzamidoacetate
SMILES [Na+].[O-]C(=O)CNC(=O)c1ccccc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H9NO3.Na/c11-8(12)6-10-9(13)7-4-2-1-3-5-7;/h1-5H,6H2,(H,10,13)(H,11,12);/q;+1/p-1
Formula Weight 201.0402
Chemical Name or Material Hippuric Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.