missing translation for 'onlineSavingsMsg'
Learn More

Hippuric acid, 98%

Product Code. 10551201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10551201 100 g Plastic bottle 100g
10541201 5 g Glass Bottle 5g
10725211 500 g Plastic bottle 500g
15159183 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10551201 Supplier Thermo Scientific Acros Supplier No. 150271000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-69-2
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00002692
InChI Key QIAFMBKCNZACKA-UHFFFAOYSA-N
Synonym hippuric acid, n-benzoylglycine, glycine, n-benzoyl, benzoylglycine, benzamidoacetic acid, benzoylaminoacetic acid, hippurate, hippursaeure, phenylcarbonylaminoacetic acid, benzamidoessigsaeure
PubChem CID 464
ChEBI CHEBI:18089
SMILES OC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 188.0°C to 191.0°C
Color White
Loss on Drying 1% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 1% max. (Benzoic acid) (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CONHCH2CO2H
Quantity 100 g
Beilstein 09,225
Fieser 01,432
Merck Index 15,4753
Solubility Information Solubility in water: 1g/250 mL cold water. Other solubilities: freely soluble in hot water,1g/100mL chloroform- 1g/400mL ether,1g/60mL amyl alcohol,freely soluble in hot ether,pract.insoluble in benzene,cs2 and petr.ether
IUPAC Name 2-benzamidoacetic acid
Formula Weight 179.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Hippuric acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.