missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hippocalcin like 4 Antibody (OTI3C3), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71923V
This item is not returnable.
View return policy
Description
Hippocalcin like 4 Monoclonal specifically detects Hippocalcin like 4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Hippocalcin like 4 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HPCAL4 | |
| Recombinant protein fragment corresponding to amino acids 2-191 of human Hippocalcin like 4 (NP_057341) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI3C3 | |
| Western Blot | |
| hippocalcin like 4, hippocalcin-like protein 4, HLP4DKFZp761G122 | |
| Mouse | |
| Affinity Purified | |
| Vision | |
| 51440.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction