missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPPI Antibody (OTI4F6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71985PE
This item is not returnable.
View return policy
Description
HIPPI Monoclonal antibody specifically detects HIPPI in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| HIPPI | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 55081 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI4F6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Dermal papilla-derived protein 8, DERP8, ESRRBL1huntingtin interacting protein-1 interacting protein, estrogen-related receptor beta like 1, Estrogen-related receptor beta-like protein 1, FLJ10147, HIPPIHIP1-interacting protein, intraflagellar transport 57 homolog (Chlamydomonas), intraflagellar transport protein 57 homolog, MHS4R2HIP1 protein interactor | |
| Full length human recombinant protein of human IFT57(NP_060480) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction