missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIPK4 Monoclonal Antibody (292CT1.4.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537528
This item is not returnable.
View return policy
Description
HIPK4 Monoclonal Antibody for Western Blot
HIPK4 is a homeodomain-interacting protein kinase and has been demonstrated to phosphorylate p53. HIPK4 encodes a member of the homeodomain interacting protein kinase (HIPK) family of proteins. While other members of this family are found throughout vertebrates, this member is present only in mammals. Compared to other members of this family, the encoded protein lacks a nuclear localization signal and a C-terminal autoinhibitory domain. The encoded protein exhibits kinase activity and may phosphorylate the tumor suppressor protein p53.
Specifications
| HIPK4 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8NE63 | |
| HIPK4 | |
| HIPK4 recombinant protein. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| Western Blot | |
| 292CT1.4.1 | |
| Unconjugated | |
| HIPK4 | |
| Gm162; Hipk4; homeodomain interacting protein kinase 4; homeodomain-interacting protein kinase 4; LOW QUALITY PROTEIN: homeodomain-interacting protein kinase 4; testicular tissue protein Li 87 | |
| Mouse | |
| Euglobulin precipitation | |
| RUO | |
| 147746 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction