Learn More
Invitrogen™ Human HIPK3 (YAK1), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic
Specifications
Specifications
| Accession Number | Q9H422 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 10114 |
| Molecular Weight (g/mol) | 49.7 kDa |
| Name | Human HIPK3 (YAK1), His Tag |
| Quantity | 10 μg |
| Regulatory Status | For research use only. Not for use in diagnostic procedures |
| Gene Alias | Androgen receptor-interacting nuclear protein kinase; ANPK; DYRK6; Fas-interacting serine/threonine-protein kinase; FIST; FIST3; HCMV-interacting protein; HIPK3; homeodomain interacting protein kinase 3; homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; mir-1902; nbak3; Nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; PKY; RP1-8L15.1; YAK1 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.