Learn More
HIPK3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Product Code. 18402068
Product Code. 18402068 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224521MFV610
Rabbit Polyclonal Antibody
HIPK3 Polyclonal antibody specifically detects HIPK3 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)