missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIPK2 Recombinant Rabbit Monoclonal Antibody (23GB2500), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HIPK2 |
|---|---|
| Clone | 23GB2500 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089780
![]() |
Invitrogen™
MA552506 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087800
|
Invitrogen™
MA552507 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIPK2 Recombinant Monoclonal antibody specifically detects HIPK2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| HIPK2 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H2X6 | |
| 28996 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2500 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110014O20Rik; B230339E18Rik; cb1009; hHIPk2; HIPK2; homeodomain interacting protein kinase 2; homeodomain-interacting protein kinase 2; Nbak1; Nuclear body-associated kinase 1; PRO0593; si:dkey-16j24.2; si:dkey-239k7.1; Sialophorin tail-associated nuclear serine/threonine-protein kinase; Stank; wu:fk56d12 | |
| A synthesized peptide derived from human HIPK2 (150-200AA). | |
| Primary | |
| -20°C | |
| HIPK2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title