missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK2 Antibody (S06-5G2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19873-100ul
This item is not returnable.
View return policy
Description
HIPK2 Monoclonal antibody specifically detects HIPK2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| HIPK2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunofluorescence | |
| S06-5G2 | |
| Unconjugated | |
| DKFZp686K02111, EC 2.7.11.1, FLJ23711, hHIPk2, homeodomain interacting protein kinase 2, homeodomain-interacting protein kinase 2, PRO0593 | |
| A synthetic peptide of human HIPK2 (Uniprot # Q9H2X6) | |
| 100 μg | |
| Apoptosis | |
| 28996 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction